Electrophoresis Chemicals, Acrylamide/ Bis-Acrylamide, G-Biosciences
Supplier: G-Biosciences
786-502 786-503 786-504 786-505 786-506 786-507 786-508 786-509
CA95043-452EA 748.48 CAD
CA95043-452 CA95043-454 CA95043-456 CA95043-458 CA95043-460 CA95043-462 CA95043-464 CA95043-466
Electrophoresis Chemicals, Acrylamide/ Bis-Acrylamide, G-Biosciences
Electrophoresis Gels Agarose and Acrylamide
G-Biosciences' Acrylamide (Electrophoresis grade) is supplied as a powder or a 40% solution in ultrapure water
G-Biosciences' Acrylamide/Bisacrylamide Solutions are available at the most common ratios (37.5:1 or 29:1) for use in protein and nucleic acid electrophoresis. The concentration is based on the total weight of both the acrylamide and bisacrylamide. Supplied as 40% solutions prepared from electrophoresis grade acrylamide and bis-acrylamide in ultra-pure water.
G-Biosciences' Acrylamide/Bisacrylamide Powders eliminate the need to weigh toxic acrylamide and bisacrylamide. Accurately pre-blended to produce a 40% (w/v) stock solution for use in protein and nucleic acid electrophoresis, these reconstituted dry powder blends are available at the most common ratios (37.5:1 or 29:1). The concentration is based on the total weight of both the acrylamide and bis-acrylamide.
Bisacrylamide (Bis (N,N’-methylenebisacrylamide)) (Electrophoresis grade) is supplied as a powder or a 2% solution in ultrapure water.