Perfect-FOCUS™ Electrophoresis Clean-Up Kit, G-Biosciences
Supplier: G-Biosciences
786-124 786-124T
CA82021-274EA 1356.67 CAD
CA82021-274 CA95029-160
Perfect-FOCUS™ Electrophoresis Clean-Up Kit, G-Biosciences
Electrophoresis Reagents Western Blotting Reagents
Perfect-FOCUS™ is designed to clean and concentrate protein samples that give poor protein spot resolution during 2D electrophoresis
Protein solutions are treated with Perfect-FOCUS™ that quantitatively precipitates the proteins, which are subsequently collected by centrifugation. The non-protein interfering agents are washed away and the protein pellet is ready to be solubilized in an appropriate sample loading buffer.
The collected protein has conductivity <50µS and is substantially free from non-protein agents, which improves spot resolution and greatly reduces spot streaking. Perfect-FOCUS™ treated protein samples are compatible with mass spectrometry analysis and show identical peptide spectra. The Perfect-FOCUS™ kit is suitable for concentrating and cleaning protein samples for isoelectric focusing (IEF) and 2D-gel electrophoresis applications.
Protein samples containing interfering agents, including ionic detergents, metal ions, substrates, substrate analogs, inhibitors, and other charged molecules, routinely result in smeared or low resolution gels.
Ordering information: Accessories not included.