Pacific Hemostasis® Prothrombin Time Reagents, Thermo Scientific
Supplier: THERMO CLINICAL CHEMISTRY
100354
100362
100356
100357
95059-796PK
1341.51
CAD
95059-796
95059-798
95059-800
95059-802
Pacific Hemostasis® Prothrombin Time Reagents, Thermo Scientific
Clinical Diagnostic Supplies
Clinical Reagents and Kits
These reagents are for the detection of acquired or congenital disorders affecting the extrinsic and common pathways
Prothrombin times are also widely accepted as a means of monitoring patients on oral anticoagulant therapy. They are compatible with automated, semi-automated, or manual systems, and have a reconstitution stability of 7 days when stored at 2 to 8°C (36 to 46°F)