Pacific Hemostasis® Activated Partial Thromboplastin Time (APTT) Reagents, Thermo Scientific
Supplier: Thermo Fisher Scientific
100402 100403
95059-804PK 477.65 CAD
95059-804 95059-806
Pacific Hemostasis® Activated Partial Thromboplastin Time (APTT) Reagents, Thermo Scientific
Clinical Diagnostic Supplies Clinical Reagents and Kits
These reagents are used to identify acquired or congenital disorders to the intrinsic and common pathways
APTT testing is widely accepted as a means of monitoring patients on oral anticoagulant therapy such as heparin. 10mL vials can complete 1000 full volume, single determinations; 4mL vials can complete 400 full volume single determinations. Controls have a two-year extended shelf-life and an open-vial stability of 30 days when stored at 2 to 8°C (36 to 46°F)