Pacific Hemostasis® SickleScreen® Assay Kit, Thermo Scientific
Supplier: THERMO CLINICAL CHEMISTRY
100258 100250
95059-772PK 2194.62 CAD
95059-772 95059-774
Pacific Hemostasis® SickleScreen® Assay Kit, Thermo Scientific
Clinical Diagnostic Supplies Clinical Reagents and Kits
Intended for use in screening for sickle cell disease and sickle cell trait, these tests are based on the differential solubility of Hemoglobin S and Hemoglobin A in a concentrated phosphate solution
The 30-test kit contains a bottle of phosphate buffer with dispensing cap and 30 pre-filled reaction vials. The 120-test kit contains four bottles of phosphate buffer and four sodium hydrosulfite vials. Control set contains two 0.5mL bottles each of positive (Hb A/Hb S) and negative (Hb A/Hb A) controls