Accessories for Gram Stain Kits, Azer Scientific
Supplier: AZER SCIENTIFIC CA
ES802 ES806 ES808 ES812-2.5G
CA89130-190CS 464.25 CAD
CA89130-190 CA89130-192 CA89130-194 CA89130-196
Accessories for Gram Stain Kits, Azer Scientific
Stains Histology Stains
These gram stain kits are used to determine the Gram reaction for microorganism identification.
  • Convenient Spout Caps
  • Comprehensive Product Insert
  • One Bottle of Each: Stabilized or Unstabilized Iodine, Crystal Violet, Decolorizer, and Safranin
Kits contain one 237mL (8oz.) bottle of each component: stabilized or unstabilized iodine, crystal violet, decolorizer, and safranin.
Detailed product insert and convenient spout caps are also included.