ICON® DS Strep A Test, Beckman Coulter

Supplier: Beckman Coulter
ICON®
395098A 395098A
CA89038-226CS 1355.8 CAD
CA89038-226 89038-226
ICON® DS Strep A Test, Beckman Coulter
Clinical Diagnostic Supplies Clinical Reagents and Kits
Simple, easy-to-use, dipstick format test features outstanding performance, even in low colony count samples, and simple-to-interpret test results.

  • Easy to use
  • No misdiagnosis
  • Accuracy 98%
  • Test time 5 min.

There is little chance for misdiagnosis, minimizing the chance of rheumatic fever or over-prescription of antibiotics.

Certifications: CLIA waived.

Ordering information: Kit contents include 25 test strips, extraction reagent A, extraction reagent B, positive control, negative control, extraction tubes, and throat swabs.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR