ICON® HP Test, Beckman Coulter
Supplier: Beckman Coulter
ICON®
395160A
CA89038-230CS 3046.04 CAD
CA89038-230
ICON® HP Test, Beckman Coulter
Clinical Diagnostic Supplies Clinical Reagents and Kits
One-step anti-H
  • One-step
  • Cassette device tests
  • Less invasive
Cassette device test aids in the diagnosis of H. pylori infection in adult patients with symptoms of gastrointestinal disorders without costly and invasive procedures.
pylori antibody test utilizes indirect solid-phase immunoassay technology for the qualitative detection of anti-Helicobacter pylori IgG antibody in human whole blood, serum, or plasma specimens.
Certifications: Whole blood is CLIA waived; serum/plasma is moderately complex.
Ordering information: Kit contents include 30 test cassettes, 35 capillary tubes, and one dropper bottle of developer solution.