ICON® Mono Test, Beckman Coulter®

Supplier: Beckman Coulter
ICON®
395100A
CA89038-228CS 1345.17 CAD
CA89038-228
ICON® Mono Test, Beckman Coulter®
Clinical Diagnostic Supplies Clinical Reagents and Kits
One-step antibody test for fast, accurate, and easy-to-read results at the point-of-care.

  • One-step
  • Easy to read
  • Accurate results
  • Sensitivity 99.9% (relative)
  • Specificity 98.8% serum/plasma (relative), 98.3% WB assay (relative)

Combines finger-stick whole blood convenience with EIA membrane technology for fast, accurate results. Uses direct solid-phase immunoassay technology for early qualitative detection of IM heterophile antibodies in whole blood, serum, or plasma. Approved for testing on all ages.

Certifications: Whole blood is CLIA waived; serum/plasma is moderately complex.

Ordering information: Kit contents include 30 test cassettes, developer solution, negative control, positive control, capillary tubes, and bulb.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR