Hemoccult II® SENSA® Fecal Occult Blood Test Systems, Beckman Coulter®
Supplier: BECKMAN COULTER(DIAGNOSTICS) CA
Rapid, convenient, and qualitative method features improved readability and higher clinical sensitivity for detecting fecal occult blood.
- Qualitative
- Sensitivity 92%
- Triple-slide
Triple-slide products are designed so patients can collect serial specimens over three days, increasing the probability of detecting hidden blood.
Certifications: CLIA waived.
Ordering information: The Dispensapak™ Plus system is supplied with 40 patient kits and two 15mL bottles of developer. Single Slides pack 10012-018 is supplied with 100 slides, 100 applicator sticks, and two 15mL bottles of developer. Single Slides 10012-020 is packaged as a case with 1000 slides, 1000 applicator sticks, and twenty 15mL bottles of developer.