Potassium iodide, ultra dry ≥99.998% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Dipotassium diiodide, Kali iodide
13676-06
13676-14
CAAA13676-06EA
104.6
CAD
CAAA13676-06
CAAA13676-14
Potassium iodide, ultra dry ≥99.998% (metals basis)
Potassium iodide
MDL: MFCD00011405
Purity: 99.998%
CAS No.: 7681-11-0
Molecular Formula: KI
Formula Weight: 166.01
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 3.12
Refractive Index: 1.677
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 681°
Boiling Point: 1330°
Merck Reference: 14,7643
MDL No.: MFCD00011405
Formula:
Kl MW: 166 g/mol Boiling Pt: 1330 Melting Pt: 686 Density: 3.12 Storage Temperature: Ambient |
MDL Number:
MFCD00011405 CAS Number: 7681-11-0 EINECS: 213-659-4 Merck Index: 14,7643 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...