Potassium iodide ≥99.0% ACS

Supplier: Thermo Scientific Chemicals

11601-22 11601-36 11601-A3
CAAA11601-22EA 104.28 CAD
CAAA11601-22 CAAA11601-36 CAAA11601-A3
Potassium iodide ≥99.0% ACS
Potassium iodide

In photographic emulsions, in analytical chemistry, as a feed additive, in spectroscopy, in infrared transmission, and in scintillation


Purity: 99.0% min CAS No.: 7681-11-0 Molecular Formula: KI Formula Weight: 166.01 Physical Form: Granular Density: 3.12 Refractive Index: 1.677 Sensitivity: Hygroscopic Odor: Odorless Melting Point: 681° Boiling Point: 1330° Solubility: Soluble in water, alcohol, acetone, and glycerol Merck Reference: 14,7643 MDL No.: MFCD00011405


In photographic emulsions, in analytical chemistry, as a feed additive, in spectroscopy, in infrared transmission, and in scintillation

Formula: Kl
Boiling Pt: 1330 °C
Melting Pt: 681 °C
Density: 3.12
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 213-659-4
Merck Index: 14,7643

Order Now

Specification Test Results

Insoluble matter 0.005%
Loss on drying at 150°C 0.2%
pH of a 5 solution 6.0-9.2 at 25°
Chloride and bromide (as Cl) 0.01%
IO3 3 ppm
PO3 0.001%
SO3 0.005%
Ba 0.002%
Ca 0.002%
MG 0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR