Potassium iodide ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Dipotassium diiodide, Kali iodide
10978-06
10978-14
10978-22
CAAA10978-06EA
97.33
CAD
CAAA10978-06
CAAA10978-14
CAAA10978-22
Potassium iodide ≥99.995% (metals basis)
Potassium iodide
MDL: MFCD00011405
Fieser: 5,327 6,227
Purity: 99.995%
CAS No.: 7681-11-0
Molecular Formula: KI
Formula Weight: 166.01
Physical Form: Crystalline
Density: 3.12
Refractive Index: 1.677
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 681°
Boiling Point: 1330°
Merck Reference: 14,7643
MDL No.: MFCD00011405
Formula:
Kl Boiling Pt: 1330 °C Melting Pt: 681 °C Density: 3.12 Storage Temperature: Ambient |
MDL Number:
MFCD00011405 CAS Number: 7681-11-0 EINECS: 213-659-4 Merck Index: 14,7643 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...