VWR® EDTA 0.5M, Biotechnology Grade
Supplier: VWR
E522-100ML
97062-836EA 80.61 CAD
97062-836
VWR® EDTA 0.5M, Biotechnology Grade
Buffers Cloning Buffers
  • DNase NONE / Fail
  • pH @25C 7.9 - 8.1
  • RNase NONE / Fail