Potassium iodide ACS
Supplier: VWR
|
Danger
|
0512-100G
0512-2.5KG
0512-500G
97061-828EA
123.27
CAD
97061-828
97061-830
97061-832
Potassium iodide ACS
Potassium iodide
Formula:
Kl Boiling Pt: 1330 °C (1013 hPa) Melting Pt: 686 °C Density: 3.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011405 CAS Number: 7681-11-0 EINECS: 231-659-4 Merck Index: 13,07727 |
Specification Test Results
Barium |
|
Calcium |
|
Chloride & Bromide |
|
Heavy Metals (as Pb) |
|
Insolubles |
|
Iodate |
|
Iron |
|
Loss on Drying |
|
Magnesium |
|
pH (5%, Water) @25°C | 6 - 9.2 |
Phosphate |
|
Purity |
|
Sodium |
|
Sulfate |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...