LabExact® Weighing Papers, Nitrogen Free
Supplier: I.W. TREMONT CO., INC.
LEW33 LEW44 LEW66
77591-702BX 25.75 CAD
77591-702 77591-704 77591-706
LabExact® Weighing Papers, Nitrogen Free
Weighing Papers
LabExact® weighing paper is an analytical purity grade non siliconized, nitrogen free paper designed for conveying powder or gelatinous paste samples to and from balances and instruments. Compatable with Kjeldahl digestion and packaged in a convenient dispensor box.
  • The material has sufficient structural integrity to resist tearing during quadrant folding or enveloping
  • The surface is highly calendared (non treated gloss) to a smoothness which resists sample hold or moisture absorption
  • Nitrogen free compositon is compatable with the Kjeldahl method
More Product Information