LabExact® Electrophoresis Blotter Cards
Supplier: I.W. TREMONT CO., INC.
LECFP1732003 LECFP1636001 LECFP1636002 LECFP1636003 LECFP1700001 LECFP1700002 LECFP1700003 LECFP1700004 LECFP1700005 LECFP1732001 LECFP1732002
77590-566BX 69.77 CAD
77590-566 77590-546 77590-548 77590-550 77590-552 77590-554 77590-556 77590-558 77590-560 77590-562 77590-564
LabExact® Electrophoresis Blotter Cards
Blotting Papers
LabExact® Electrophoresis blotter cards are highly consistent general purpose electrophoresis blotters that are naturally hydrophilic with no additional surfactants or additives necessary to yield high performance surface absorption rates and migration speeds.
  • Smooth surface, bright white pure cotton cellulose paper promotes clean transfers and blots
  • Material is manufactured from carefully selected, USA sourced pure alpha cellulose fiber which is fully oxygen bleached and resin free
  • Highly absorbent and very rapid migration
  • Rapid surface absorption wicks moisture off membrane and gel pads quickly and reliably
  • Manufactured in the USA
The even density and high grade formation yields a very consistent and reproducible migration in both MD and CD.
Naturally hydrophilic and hydroscopic with no additional surfactants or additives necessary to yield high performance surface absorption rates and migration speeds and free of additives which can interfere with the membrane indication and transfer process.
Caution: For research only, not a medical device PCR component.