LabExact® High Purity CA Fibrous Protein Syringe Filters
Supplier: I.W. TREMONT CO., INC.
LEES10751 LEES10752
77591-136PK 389.77 CAD
77591-136 77591-138
LabExact® High Purity CA Fibrous Protein Syringe Filters
Filters Syringe Filters
LabExact® Fibrous Cellulose Acetate depth filter media syringe filters with 1.0 µm retention demonstrate similar advantages as its Cellulose Acetate surface membrane counterparts such as surface bonding, high purity and biocompatibility in addition this depth filter media has a high surface area to mass ratio and natural high loading capacity.
  • It has an inherent charge that initially has high protein binding but upon loading, releases easily and completely for high recovery rates
  • Filters are ultrasonically bonded for reliability and the filter media type is clearly inscribed on barrel
  • The filters feature Luer-Lok™ and pressure fitting
  • Excellent unit-to-unit and lot-to-lot reproducibility and consistency
  • The concentric funnel shaped barrel increases volume and decreases back pressure