LabExact® PVDF Syringe Filters with Glass fiber Prefilter
Supplier: I.W. TREMONT CO., INC.
LEES10219 LEES10220 LEES10221 LEES10222
77591-034PK 386.33 CAD
77591-034 77591-036 77591-038 77591-040
LabExact® PVDF Syringe Filters with Glass fiber Prefilter
Filters Syringe Filters
LabExact® disposable non-sterile PVDF hydrophobic filtration media syringe filters feature stability and hydrophobic interactions similar to PTFE with a 1 µm glass microfiber prefilter that raises absorption capacity and loading.
  • Recommended especially for filtration of water-soluble oligomers and polymers, e.g. protein solutions
  • Can also be used for the filtration of hydrophobic substances in different organic solvents
Syringe filters feature Luer-Lok™ and pressure fitting. They are ultrasonically bonded for reliability and the filter media type isMore Product Information