LabExact® Cellulose Acetate Syringe Filters
Supplier: I.W. TREMONT CO., INC.
LEES10081 LEES10082 LEES10083 LEES10084 LEES10611 LEES10612
77590-986PK 202.7 CAD
77590-986 77590-988 77590-990 77590-992 77591-118 77591-120
LabExact® Cellulose Acetate Syringe Filters
Filters Syringe Filters
LabExact® disposable non-sterile Cellulose Acetate hydrophilic syringe filters have very low binding capacity for proteins and used for filtration and clarification of biological fluids.
  • Cellulose acetate is nitrate-free making it suitable for groundwater and EPA ASTM’s
  • Use for filtration under sterile conditions or filtration and clarification of biological fluids, sterilization of media additives and serum and sample preparation of aqueous or protein-based HPLC solutions
  • Use for applications requiring maximum sample recovery
  • Designed to sterilize and remove particulates from even the most viscous proteinaceous samples
  • Use with a glass fiber prefilter to filter tissues culture media
Filters feature Luer-Lok™ and pressure fitting. They are ultrasonically bonded for reliability and the filter media type is clearly inscribed on barrel. Excellent unit-to-unit and lot-to-lot reproducibility and consistency. The concentric funnel shaped barrel increases volume and decreases back pressure.