LabExact® Tri-Layer Syringe Filters with GF/PP Prefilters, Non Sterile
Supplier: I.W. TREMONT CO., INC.
LEES10405 LEES10406 LEES10410 LEES10411 LEES10400 LEES10401
77591-086PK 560.41 CAD
77591-086 77591-088 77591-090 77591-092 77591-082 77591-084
LabExact® Tri-Layer Syringe Filters with GF/PP Prefilters, Non Sterile
Filters Syringe Filters
LabExact® disposable non sterile tri-layer (PTFE, nylon/polyamide and PVDF) filtration media syringe filters with glass microfiber and polypropylene prefilter offer increased absorption capacity and loading and are commonly used for general laboratory filtration and filtration of HPLC and GC samples prior to injection in analyzer.
  • Available with Nylon or PVDF or PTFE membrane with glass microfiber and polypropylene prefilter that raises absorption capacity and loading
  • Suitable for filtration of aqueous and organic-aqueous polar medium liquids and
More Product Information