LabExact® MCE Syringe Filters, Non Sterile
Supplier: I.W. TREMONT CO., INC.
LEES10098 LEES10099 LEES10100 LEES10101
77590-994PK 173.33 CAD
77590-994 77590-996 77590-998 77591-000
LabExact® MCE Syringe Filters, Non Sterile
Filters Syringe Filters
LabExact® disposable non sterile MCE hydrophilic filtration media syringe filters are recommended for all filtrations in aqueous or polar media, bacteria filtration.
  • Hydrophilic filteration media
  • Ideal when maximum protein recovery in filtrate is crucial
  • Used for filtration and inspection of gas, oil and alcohol particulate and bacteria
  • When combined with glass microfiber prefilter, it is ideal for tissue culture media and sensitive biological samples
  • Feature Luer-Lok™ and pressure fitting
  • They are ultrasonically bonded for reliability and the filter media type is clearly inscribed on barrel
Excellent unit-to-unit and lot-to-lot reproducibility and consistency. The concentric funnel shaped barrel increases volume and decreases back pressure.