LabExact® Trupor® Nylon High Flow, Low Back Pressure Filter Membranes
Supplier: I.W. TREMONT CO., INC.
LETNY045090 LETNY045142 LETNY020025 LETNY020047 LETNY020090 LETNY020142 LETNY045025 LETNY045047
77591-606BX 609.75 CAD
77591-606 77591-608 77591-594 77591-596 77591-598 77591-600 77591-602 77591-604
LabExact® Trupor® Nylon High Flow, Low Back Pressure Filter Membranes
Filters Membrane Filters
H&V Trupor® Nylon membranes utilize nanoscale technology in a composite design to achieve precisely regulated porosities with higher loading capacity and faster flow. No adhesives are utilized, which significantly reduces trace extractables.
  • No additives are utilized, which significantly reduces trace extractables
  • Trupor® membrane filters demonstrate back pressure reduction by >50% versus comparable porosity cast membranes
  • Trupor® Nylon membrane filters demonstrate loading capacity >40% versus comparable porosity cast membrane
  • Larger sample volumes can be prepared with one filter to reduce cost in sample preparation, increase workflow, and decrease waste disposal
  • Autoclavable
The advantages of lower back pressure and greater loading capacities do not compromise precision pore structure. Trupor® membrane filter retention testing demonstrates equal or greater consistency to many leading polymeric cast film membranes on the market today. Feel secure that critical samples are being separated to the target retention consistently from device-to-device and lot-to-lot.
Packaging: Box of 100.