Human IR-B Luciferase Reporter Assay Kit
Supplier: INDIGO BIOSCIENCES INC MS
IB28001 IB28002
104087-916EA 1600 CAD
104087-916 104087-918
Human IR-B Luciferase Reporter Assay Kit
Assays Enzyme Assays
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO's insulin receptor, isoform B (IR-B) reporter assay allows you to quickly screen test samples to quantify functional activity, either agonist or antagonist, they may exert against IR-B.
  • Includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor
  • Kit includes reporter cells, optimized media for use during cell thawing and in diluting test samples, reference agonist, luciferase detection reagent, cell culture-ready assay plate, and detailed protocol
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot (product quality data available upon request)
This is an all-inclusive cell-based luciferase reporter assay kit targeting the human insulin receptor, isoform B (IR-B). INDIGO's IR-B reporter assay utilizes proprietary mammalian cells that have been engineered to provide constitutive expression of IR-B. In addition to IR-B reporter cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, a reference agonist, luciferase fetection reagent, and a cell culture-ready assay plate. The principal application of this assay is in the screening of test samples to quantify any functional activity, either agonist or antagonist, that they may exert against IR-B. This kit provides researchers with: Clear, reproducible results, exceptional cell viability post-thaw. Consistent results lot to lot.
Ordering information: Kits are all-inclusive assay systems featuring live cells.
Delivery information: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at −80 °C. Do not store in liquid nitrogen.