Human G-CSFR Luciferase Reporter Assay Kit
Supplier: INDIGO BIOSCIENCES INC MS
IB30101 IB30102
104087-928EA 1606.15 CAD
104087-928 104087-930
Human G-CSFR Luciferase Reporter Assay Kit
Assays Enzyme Assays
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO's Granulocyte Colony-Stimulating Factor Receptor reporter assay allows you to quickly screen test samples to quantify functional activity, either agonist or antagonist, they may exert against G-CSFR.
  • Includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor
  • Kit includes reporter cells, optimized media for use during cell thawing and in diluting test samples, reference agonist, luciferase detection reagent, cell culture-ready assay plate, and detailed protocol
  • Exceptional cell viability post-thaw
  • Consistent results lot to lot (product quality data available upon request)
This is an all-inclusive cell-based luciferase reporter assay kit targeting the Granulocyte Colony-Stimulating Factor Receptor (G-CSFR). INDIGO's G-CSFR reporter assay utilizes proprietary mammalian cells that have been engineered to provide constitutive expression of the G-CSFR. In addition to G-CSFR reporter cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, a reference agonist, Luciferase detection reagent, and a cell culture-ready assay plate. The principal application of this assay is in the screening of test samples to quantify any functional activity, either agonist or antagonist, that they may exert against G-CSFR. This kit provides researchers with: Clear, reproducible results, exceptional cell viability post-thaw. Consistent results lot to lot.
Ordering information: Kits are all-inclusive assay systems featuring live cells.
Delivery information: Kits are shipped via overnight delivery on dry ice. Due to extreme temperature-sensitivity of the live cells, kits must be stored at −80 °C. Do not store in liquid nitrogen.