OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Supplier: Sekisui Diagnostics
OSOM®
147 149 149 147
CA-71001-042CS 5175.13 CAD
71001-042 CA-71001-042 71001-050 CA71001-050
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
These convenient test kits provide easy-to-read, two-color results in as little as five minutes
Studies have shown that the sensitivity of Ultra Strep A is statistically equivalent to single swab culture. Kit contents maintain a room temperature shelf life of 15 months from the date of manufacture.
Certifications: CLIA-waived.
Ordering information: The 50-test kit includes two additional tests for external QC testing.