OSOM® Mono Test Kit, Sekisui Diagnostics

Supplier: Sekisui Diagnostics
OSOM®
145 145
CA71001-040CS 3572.6 CAD
CA71001-040 71001-040
OSOM® Mono Test Kit, Sekisui Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
Detects heterophile antibodies to EBV (mononucleosis) in serum, plasma, or whole blood.

Easy-to-read two color results in five minutes or less (Sensitivity 100%, Specificity > 99%).

For use in ages 4 and up. The Mono test maintains a room temperature shelf life of 12 months from the date of manufacture.

Certifications: CLIA-waived for whole blood. CE marked.

Ordering information: Kit includes two additional tests for external QC testing (25-test kit includes 27 tests).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR