GelBond® Film
Supplier: Lonza
53734 53740 53745 53746 53748 53749 53750 53759 53761 53780 54711 54723 54727 54729 54731 54733 54746
CA12001-936EA 785.54 CAD
CA12001-936 CA12001-938 CA12001-940 CA12001-942 CA12001-944 CA12001-946 CA12001-948 CA12001-950 CA12001-954 CA12001-956 CA12001-958 CA12001-960 CA12001-962 CA12001-964 CA12001-966 CA12001-968 CA12001-972
GelBond® Film
Seals Microplate Seals
GelBond Film® is a transparent, flexible polyester film designed to support agarose gels. Gels cast on GelBond® Film remain permanently attached to the film through electrophoresis or immunodiffusion and all subsequent fixing, staining, destaining, and drying procedures.
  • Will not shrink or crack
  • Easier to handle during staining, destaining, and drying when supported
  • Durable and convenient
Gels cast on the film retain their original dimensions during staining and after drying. The orientation can be recorded directly on the film prior to casting to eliminate confusion that can occur during staining and drying procedures.
GelBond® film is designed to be used with agarose gels. GelBond PAG film is designed to be used with polyacrylamide or MDE® gels.
Caution: Polyester films will not transmit light less than 310 nm and will fluoresce at higher wavelengths.