BD Hycheck™ Paddles for Monitoring Production Environments, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
290001 290005 290002 290003 290004 290006 290007 290003
CA90004-100PK 87.56 CAD
CA90004-100 CA90004-108 CA90004-102 CA90004-104 CA90004-106 CA90004-110 CA90004-112 CA89200-310
BD Hycheck™ Paddles for Monitoring Production Environments, BD Diagnostics
Microbiology Test Systems Microorganism Tests
BD HYcheck™ is a hygiene contact slide used to assess the microbiological contamination of surfaces or fluids
The double-sided slide features a hinged paddle that bends for easy sampling.
Ordering information: Store at 2 to 15°C (36 to 59°F). Multiple media combinations are available; contact your Avantor sales representative for details.