BD BBL™ Thioglycollate Medium, Prepared in Tubes, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
221200 221788
CA-90001-264PK 184.65 CAD
CA-90001-264 CA-90001-522
BD BBL™ Thioglycollate Medium, Prepared in Tubes, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Media for the isolation and identification of microorganisms.
  • Tubes are made of borosilicate glass
  • Long-necked, rubber-lined screw caps prevent contamination
  • Larger-sized tubes with wider diameters permit easy inoculation
  • Securely fitted screw caps prevent loss of moisture
Media in Tubes are ideal for cultures requiring prolonged incubation, since the caps can be closed tightly to prevent dehydration and subsequent inhibition of growth. Caps contain a resilient rubber liner that resists displacement even during autoclaving.