OSOM® hCG Test Kits, Sekisui Diagnostics
Supplier: SEKISUI DIAGNOSTIC CA
101 102W 124 124
CA71001-030CS 3580.6 CAD
CA71001-030 CA71001-033 CA71001-044 71001-044
OSOM® hCG Test Kits, Sekisui Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
This variety of hCG testing kits and procedures allows users to select the method that best suits the facility in use.
The hCG Urine Test Kit offers an economic and convenient procedure that utilizes a color immunochromatographic dipstick testing method. Easy-to-read two-color results are available in as little as one minute. Test is 99% accurate. The kit includes 50 tests, and maintains an 18-month shelf life from the date of manufacture when stored at room temperature. Sensivity 25 mIU/mL. CLIA-waived.
The Urine Card Pregnancy Test Kit is a traditional physician's office test that utilizes a solid phase, sandwich-format immunochromatographic assay testing method. Tests provide easy-to-read black and white results in 3 minutes. The kit includes 25 tests, and maintains a 24-month shelf life from the date of manufacture when stored at room temperature. Sensivity 20 mIU/mL. CLIA-waived.
The hCG Combo Test Kit is a rapid immunoassay for the qualitative detection of hCG in urine or serum. Designed for use in physician's offices or clinical laboratories, the test utilizes a solid phase, sandwich-format immunochromatographic assay testing method. Urine samples provide results in 3 minutes; serum samples in 5 minutes. The kit includes 25 tests, and maintains a 24-month shelf life from the date of manufacture when stored at room temperature. Sensivity 10 mIU/mL for Serum; 20 mIU/mL for Urine. CLIA-waived for urine.
Urine and serum control sets are sold separately. Urine control set includes one negative and two levels of positive urine controls. Serum control set includes one negative and one positive serum control. Control sets require refrigerated storage.