Difco™ L-Arabinose/Dextrose/Cellobiose, BD Biosciences
Supplier: BD CANADA MICROBIOLOGY CA
215520 215510 215530
CA90000-906EA 1089.42 CAD
CA90000-906 CA90000-904 CA90000-908 CA90000-918 CA90000-920
Difco™ L-Arabinose/Dextrose/Cellobiose, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Media formulations in dehydrated form is for the convenience of the laboratorian and bioprocessing specialist.
  • Consistent, high-quality performance
  • Conveniently dehydrated
The individual ingredients and the final dehydrated product are carefully quality-controlled to assure consistent, high quality performance and obviate the need for media to be prepared in the laboratory from raw materials. Dehydrated culture media may be offered as 100 g and 500 g bottles, 2 kg and 10 kg containers.
Glucose, D (+), anhydrous, for incorporation into microbiological culture media as a source of energy for bacteria and for the determination of fermentation reactions. It is usually employed at a concentration of 0.5 to 1.0% in fermentation media.
Order Now
SPECIFICATIONS
Environmentally Preferable