BD BBL™ Trypticase™ Soy Agar with Lecithin and Polysorbate 80 Finger Dab™ Plates, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
292271
CA90004-200PK 242.5 CAD
CA90004-200 CA-90007-752
BD BBL™ Trypticase™ Soy Agar with Lecithin and Polysorbate 80 Finger Dab™ Plates, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
BD BBL™ prepared agar plates in sterile packs are plated media in gamma-irradiated packs that are validated to performance and sterility according to AAMI (Association for the Advancement of Medical Instrumentation) guidelines
  • For critical environments
  • Validated to 10-6 SAL
  • Sterile inner wrapping
Outer Tyvek/polyethylene wrapping minimizes the risk of false contamination, provides fiber-free bacterial barrier and allows for gas exchange. To use, remove the outer wrapping in the non-critical environment, then remove the sterile inner wrapping in the clean area; the included bag may be used to transport plates from your critical environment. Storage temperature: 2 to 8 °C.
BD Finger Dab™ agar plates for sampling gloved hands.
Ordering information: Each pack or case contains a signed, lot-specific Certificate of Analysis.