Coventry™ Wrapped Foam Swabs, ITW Chemtronics®
Supplier: ITW Chemtronics
20050 20080
15145-612PK 708.39 CAD
15145-612 76240-050
Coventry™ Wrapped Foam Swabs, ITW Chemtronics®
Swabs Swabs for Cleaning
This durable adhesive-free swab are designed for precise applications and provides good particle entrapment and works well as a adhesive, lubricant and solvent applicators.
  • Wicking action pulls solvent from surfaces
  • Fiberless construction does not generate loose fibers or particles
  • No adhesives or binders are used in the construction
  • Stronger head than other foam swabs
Its small tip is ideal for use in tight tolerance areas. The swab has a wrapped, open-cell polyurethane foam head on a nylon or wood handle. Wrapped foam is durable and delivers superior abrasion resistance for the removal of tough contaminants.