Stain Kits, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
212520 212521 212522
CA-90008-884EA 248.66 CAD
CA-90008-884 CA-90008-886 CA-90008-888
Stain Kits, BD Diagnostics
Stains Microbiology Stains
These kits include all the necessary components for categorizing bacteria as gram-negative or gram-positive, a definitive characteristic in the organism identification process.
Fluorescent stain kit 90008-886 enables the observation of smears at a lower magnification, increasing the field of view and decreasing examination time. Kit includes one 250mL bottle each of the following: TB Auramine-Rhodamine T, TB Decolorizer TM, and TB Potassium Permanganate.
TB stain kit K 90008-888 includes one 250mL bottle each of TB Carbolfuschin KF, TB Decolorizer, and TB Brilliant Green K.
TB stain kit ZN 90008-884 includes one 250mL bottle each of TB Carbolfuschin ZN, TB Decolorizer, and TB Methylene Blue.
Ordering information: All stain kits come conveniently packaged in a compact tray and bottles feature no-drip “keep clean” cap spouts for dispensing.