Caspase-3 Substrate VII, Fluorogenic, Calbiochem®
Supplier: MilliporeSigma
264151-1MG 264151-5MG
CA80051-372EA 358.76 CAD
CA80051-372 CA80051-374
Caspase-3 Substrate VII, Fluorogenic, Calbiochem®
Electrophoresis Reagents Western Blotting Reagents
Fluorogenic caspase-3 substrate
Also acts as a substrate for granzyme-B processed ICE-LAP3, caspase-6, caspase-7, caspase-8, and caspase-10. Excitation maximum: ~400nm; emission maximum: ~505nm.