Caspase-3 Substrate II, Fluorogenic, Calbiochem®
Supplier: MilliporeSigma
235425-1MG
235425-5MG
CA80051-242EA
318.19
CAD
CA80051-242
CA80051-244
Caspase-3 Substrate II, Fluorogenic, Calbiochem®
Electrophoresis Reagents
Western Blotting Reagents
Fluorogenic caspase-3 substrate
Cleavage of this substrate by caspase-3 shows Michaelis-Menten kinetics (Km=9.7±1.0µM). Also a substrate for caspase-6, caspase-7, caspase-8, and caspase-10. Excitation maximum: ~365–380nm; emission maximum: ~430–460nm.