BD BBL™ and BD Difco™ Gram Stain Kit Components, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
Each Gram crystal violet solution contains approximately 0.3% crystal violet aqueous alcohol solution
Gram stain QC slides are conventional 25x75mm (1x3") microscope slides imprinted with ten squares. One square contains gram-positive and gram-negative control organisms Staphylococcus aureus and E. coli and the remaining nine squares are available for staining test isolates.
Each Gram decolorizer solution contains a 3:1 ratio of ethanol to acetone to assure even and consistent decolorization. Each Gram iodine solution (stabilized) contains an organically stabilized iodine-polyvinylpyrrolidone (PVP), which retains a safe and constant level of unoxidized iodine to maintain effectiveness as a mordant for at least 15 months. Gram iodine (unstabilized) is designed to prepare a traditional Gram’s working solution without requiring a breakage of ampules. Safranin is a mixture of dimethyl and trimethyl and is most commonly used for counterstaining nuclei red. Basic Fuchsin is a mixture of Pararosanilin, Rosanilin, and Magenta II.
Ordering information: All containers feature no-drip cap spout. Kits are packaged in convenient compact trays.