UltraFit™ Nylon Glove Liners, Erie Scientific (Thermo Scientific)
Supplier: Thermo Scientific
51002-12-001 51004-12-001 51001-12-001 51003-12-001
56613-472CS 1212.11 CAD
56613-472 56613-474 56614-284 56614-288
UltraFit™ Nylon Glove Liners, Erie Scientific (Thermo Scientific)
Gloves
Protect extra-sensitive hands from irritation and occlusion of the skin associated with frequent gloving with Thermo Scientific™ UltraFIT™ Nylon glove liners.
  • These liners are are pre-shrunk for multiple laundering
  • Form-fitting design for a skin-tight fit
  • Ultra-thin fabric for optimum dexterity and sense of touch
  • Extra layer of insulation when working in cooler areas
  • Seamless fingertips
  • Cuffs are color coded for easy sorting
  • Nonlinting and washable
These glove liners offer added protection to individuals whose hands are extra-sensitive to theMore Product Information
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable