Potassium iodide ≥99.0%, granular, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

PX1507-3 PX1507-7 PX1507-30 PX1507-1 PX1507-7 PX1507-5 PX1507-5 PX1507-3 PX1507-30 PX1507-1
CAPX1507-7EA 3701.99 CAD
CAPX1507-7 CAPX1507-5 EM-PX1507-5 EM-PX1507-7 CA-PX1507-30 EM-PX1507-3 EM-PX1507-30 CAPX1507-3 EMPX1507-1 CAPX1507-1
Potassium iodide ≥99.0%, granular, GR ACS, Supelco®
Potassium iodide

Meets ACS Specifications

Formula: Kl
MW: 166 g/mol
Boiling Pt: 1325 °C (1013 hPa)
Melting Pt: 681 °C (lit.)
MDL Number: MFCD00011405
CAS Number: 7681-11-0
Merck Index: 13,07727

Order Now

Specification Test Results

Assay 99.0% min
Barium (Ba) 0.002% max
Calcium (Ca) 0.002% max
Chloride and bromide (as Cl) 0.01% max
Color White
Form Granules [~18 mesh]
Heavy metals (as Pb) 5ppm max
Insoluble matter 0.005% max
Iodate (IO3) 3ppm max
Iron (Fe) 3ppm max
Loss on drying (150°C) 0.2% max
Magnesium (Mg) 0.001% max
pH of a 5% solution at 25°C 6.0-9.2
Phosphate (PO4) 0.001% max
Sodium (Na) 0.005% max
Sulfate (SO4) 0.005% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR