Anti-P2RX7 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-22876R
77439-726EA 610.28 CAD
77439-726
Anti-P2RX7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: P2RX7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR