Anti-TEFF1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-17320R
77438-190EA 601.4 CAD
77438-190
Anti-TEFF1 Rabbit Polyclonal Antibody
Antibodies
This locus represents naturally occurring read-through transcription from the neighboring MSANTD3 (Myb/SANT-like DNA-binding domain containing 3) and TMEFF1 (transmembrane protein with EGF-like and two follistatin-like domains 1) genes. The read-through transcript encodes a fusion protein that shares sequence identity with the products of each individual gene. [provided by RefSeq, Aug 2013]

Type: Primary
Antigen: TEFF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR