Anti-C19orf57 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-13791R
77440-442EA 610.28 CAD
77440-442
Anti-C19orf57 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: C19orf57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR