AASTY 11-45

Supplier: CUBE BIOTECH

18255 18256 18257
77392-532PK 532.62 CAD
77392-532 77392-534 77392-536
AASTY 11-45
AASTY 11-45

AASTYs (Acrylic acid-co-styrenes) - like AASTY 11-45 - are highly alternating copolymers, well-suited for the generation of native lipid nanodiscs. They are a 2022 novel developed series for membrane protein solubilization & stabilization. AASTY 11-45 gets its name from its molecular weight and Acrylic Acid : Styrene Ratio. These varying ratios of acrylic acid to styrene contribute to the hydrophilic properties of our AASTYs. In general lighter AASTYs, like 6-45 tend to be more aggressive, while heavier AASTYs, such as 11-45 show higher thermodynamic stability.

The exact composition of AASTY copolymers shows different extraction efficiency, depending on the lipid composition of the lipid bilayers being formulated into nanodiscs. As AASTY is made by controlled radical polymerization techniques, the dispersity of polymer molecular weight distribution is low, and the molecular weights are controlled. This means that excess AASTY copolymer can be removed by dialysis after nanodisc formation. Based on previous findings on SMA, it is the expectation that AASTY of different molecular weights will display different rates of nanodisc formation, extraction efficacy, and stability of resulting nanodiscs.

Every membrane protein solubilization needs to undergo a screening process before. The characteristic phospholipid environment surrounding the different membrane proteins in question performs differently well with each polymer. To support you in this process, we offer a handy Screening Kit for AASTYs to test them all. Additionally, we recommend the two following publications if you would like to get further information: Smith et al. 2020 & Timcenko et al. 2022


State: Lyophilized Powder, to be solved with water
Molecular weight: 10-11 kDa
Buffer: HEPES
pH after solving: 7.5
Divalent cationic tolerance: 6 mM
Acrylic Acid to Styrene Ratio:45% to 55%


Caution: Shipping Temperature: Ambient temperature. Storage of lyophilized copolymer: -20°C for several years. Storage of dissolved copolymer: 2-8°C for several day.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR