SMALP 140-I

Supplier: CUBE BIOTECH

18230 18231 18232
77392-520PK 532.62 CAD
77392-520 77392-522 77392-524
SMALP 140-I
SMALP 140-I

The SMALP products have been developed for biomedical purposes, in particular, for the solubilization of (biological) lipid membranes and isolation & purification of integral membrane proteins.
After solving the SMA + buffer powder in water, the product is ready-to-use. It can primarily used for all cell membrane hosts, including bacteria, yeast, and human cells. After successful solubilization of the membrane protein, the protein can be purified using affinity chromatography. For membrane protein purification we recommend using the Rho1D4-tag. Cube Biotech offers matching products for this purpose.


State: Lyophilized Powder, to be solved with water
Polymer styrene-to-maleic anhydride ratio: 1.4:1
Molecular weight: 5000 Da
pH after solving: 7.5
Solubility: 45 %
Divalent cationic tolerance: < 5 mM
Color: Amber
Odor: Odorless
Freeze-thaw stable?: Yes


Caution: Shipping Temperature: Ambient temperature. Storage of lyophilized copolymer: -20°C for several years. Storage of dissolved copolymer: 2-8°C for several day.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR