SMALP 200

Supplier: CUBE BIOTECH

18210 18211 18212
77392-508PK 532.62 CAD
77392-508 77392-510 77392-512
SMALP 200
SMALP 200

The SMALP products have been developed for biomedical purposes, in particular, for the solubilization of (biological) lipid membranes and isolation & purification of integral membrane proteins.
After solving the SMA + buffer powder in water, the product is ready-to-use. It can primarily used for all cell membrane hosts, including bacteria, yeast, and human cells. After successful solubilization of the membrane protein, the protein can be purified using affinity chromatography. For membrane protein purification we recommend using the Rho1D4-tag. Cube Biotech offers matching products for this purpose.


State: Lyophilized Powder, to be solved with water
Polymer styrene-to-maleic anhydride ratio: 2:1
Molecular weight: 6,500 Da
pH after solving: 7.5
Solubility: 40 %
Divalent cationic tolerance: < 5 mM
Color: Yellowsh
Odor: Odorless
Freeze-thaw stable?: Yes
Other noteworthy feature: Most commonly used SMA


Caution: Shipping Temperature: Ambient temperature. Storage of lyophilized copolymer: -20°C for several years. Storage of dissolved copolymer: 2-8°C for several day.

CAS Number: 25736-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR