3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS)

Supplier: CUBE BIOTECH

16080 16081 16082
77392-418PK 166.03 CAD
77392-418 77392-420 77392-422
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS)
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)

CHAPS is a zwitterionic detergent and its name is an abbreviation for 3-[(3-cholamidopropyl)dimethylammonio]-1-propanesulfonate. Detergents are the most commonly used substances for membrane protein stabilization. Before you use a specific detergent for a new membrane protein you must screen for a matching one.


-

Formula: C₃₂H₅₈N₂O₇S
MDL Number: MFCD00081083
CAS Number: 75621-03-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3aa5ef7e0e0d192f4ba2ae106b6e5c5d2bb6dcaf&vid=f8f5aaf7c83475e0bccc5e78fc6fcfd7642a48e9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR