n-Tridecyl-β-D-maltoside ≥99.5%, Crystallization grade

Supplier: CUBE BIOTECH

16075 16076 16077 16074
77392-412PK 1642.85 CAD
77392-412 77392-414 77392-416 77392-410
n-Tridecyl-β-D-maltoside ≥99.5%, Crystallization grade
n-Tridecyl-β-D-maltoside

Mild, non-ionic detergents such as Tridecylmaltoside are well suited for the solubilization, stabilization, and purification of membrane proteins. For screening of the best-suited detergent for a given application, we offer maltosides with chain lengths from C-9 to C-13.


Molecular Weight: 468.41 g/mol
Form: White crystalline powder
CMC (in H2O): 6 mM
Solubility: H2O: 10 mg/ml, clear, colorless
Hill notation: C25H48O11

Formula: C₂₅H₄₈O₁₁
MDL Number: MFCD08062386
CAS Number: 93911-12-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69d82e863260b6783d9f91200d7f730dc4e75401&vid=c160805eca35193bcb27339ce14c0a32a5ae8998&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR