N-Undecyl Beta-maltoside ≥99.5%, Crystallization grade

Supplier: CUBE BIOTECH

Synonyms: UDM

16070 16071 16072
77392-404PK 696.91 CAD
77392-404 77392-406 77392-408
N-Undecyl Beta-maltoside ≥99.5%, Crystallization grade
N-Undecyl Beta-maltoside

Mild, non-ionic detergents such as Undecylmaltoside are well suited for the solubilization, stabilization, and purification of membrane proteins. For screening of the best-suited detergent for a given application, we offer maltosides with chain lengths from C-9 to C-13.


Molecular Weight: 496.6 g/mol
Form: White crystalline powder
CMC (in H2O): 0.59 mM

MW: 496.6 g/mol
CAS Number: 253678-67-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=481bbb6afdc223b421edd05dc064b45a89f9d1e6&vid=712217a8d69ca9f56e9c6db07beac4230a0500b7&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6588e913ced4c9abab60b2f4355dee661461b0&vid=67ae7aeb6c0fa940381ce6063dcb6b73969eda47&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR