N-Nonylphosphocholine ≥99.5%, Crystallization grade

Supplier: CUBE BIOTECH

16049 16050 16051 16055
77392-380EA 389.98 CAD
77392-380 77392-382 77392-384 77392-386
N-Nonylphosphocholine ≥99.5%, Crystallization grade
N-Nonylphosphocholine

Phosphocholines, or Fos-cholines, are a novel class of detergents that is well suited for the solubilization, stabilization, and purification of membrane proteins. For screening of the best-suited detergent for a given application, we offer Fos-cholines with chain lengths from C-9 to C-16.


Molecular Weight: 309.4 g/mol
Form: White crystalline powder
CMC (in H2O): 32 mM (0.99%)
Synonyms are: Nonylphosphocholine, Fos-9

CAS Number: 253678-64-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=481bbb6afdc223b421edd05dc064b45a89f9d1e6&vid=712217a8d69ca9f56e9c6db07beac4230a0500b7&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=373685d62f1bc65e5e623cb6336f28d91dbd03eb&vid=4b0bd4cdce6ab54db18825f9afcb3072fa3429b8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR